Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143082 Supplier Invitrogen™ Supplier No. PA5143082
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element, an octameric palindrome. The protein interacts with host cell factor C1, which also associates with the herpes simplex virus (HSV) protein VP16 that induces transcription of HSV immediate-early genes. This protein and VP16 both bind to the same site on host cell factor C1. It is thought that the interaction between this protein and host cell factor C1 plays a role in the establishment of latency during HSV infection. An additional transcript variant has been identified, but its biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CREB3
Gene Accession No. O43889
Gene Alias AU044960; AW538053; basic leucine zipper protein; C80076; cAMP responsive element binding protein 3; cAMP-responsive element-binding protein 3; CREB3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; cyclic AMP-responsive element-binding protein 3; leucin zipper proitein; Leucine zipper protein; Luman; LZIP; LZIP-1; LZIP-2; N-terminal Luman; Processed cyclic AMP-responsive element-binding protein 3; sLZIP; small leucine zipper protein; transcription factor LZIP; transcription factor LZIP-alpha
Gene Symbols CREB3
Host Species Goat
Immunogen Peptide with sequence C-HHDHTYSLPRET (aa 76-87).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10488
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.