Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116414
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116414 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116414 Supplier Invitrogen™ Supplier No. PA5116414
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

LZK Polyclonal Antibody for Western Blot, ICC/IF

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine-like kinase (TLK) group consists of 40 tyrosine and serine-threonine kinases such as MLK (mixed-lineage kinase), LISK (LIMK/TESK), IRAK (interleukin-1 receptor-associated kinase), Raf, RIPK (receptor-interacting protein kinase), and STRK (activin and TGF-beta receptors) families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Clone 810CT26.6.1.1
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAP3K13
Gene Accession No. O43283
Gene Alias C130026N12Rik; Leucine zipper-bearing kinase; LZK; Map3k13; MEKK13; mitogen activated protein kinase kinase kinase 13; mitogen-activated protein kinase kinase kinase 13; Mixed lineage kinase; MLK
Gene Symbols MAP3K13
Host Species Mouse
Immunogen purified His-tagged MAP3K13 protein fragment.
Purification Method Euglobulin precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.