Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LZTFL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118040
This item is not returnable.
View return policy
Description
LZTFL1 Polyclonal Antibody for Western Blot, IHC (P), IP
LZTFL1, also known as Leucine Zipper Transcription Factor-Like 1, is situated on the chromosomal region 3p21.3, which is frequently deleted in various cancers, including kidney cancer. It acts as a tumor suppressor by destabilizing AKT, a key protein in cell growth pathways, thus inhibiting tumor cell growth. Additionally, LZTFL1 has been implicated in the epithelial-mesenchymal transition in COVID-19, suggesting a role in viral response pathways. It is associated with Bardet-Biedl syndrome (BBS) and interacts with IFT27, a regulator of sperm flagellum formation, affecting spermatogenesis and male fertility. Knockout models show a phenotype of obesity, retinal degeneration, and abnormal cilia development, highlighting its role in normal ciliary function. The gene has two transcript isoforms and is part of a transcriptional map covering 250 kb, making it a subject of extensive genetic research.
Specifications
LZTFL1 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9NQ48 | |
Lztfl1 | |
E. coli-derived Human LZTFL1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Lztfl1 | |
5530402H04Rik; 6130400H19Rik; AI414725; AW048545; BBS17; fb53f12; leucine zipper transcription factor like 1; leucine zipper transcription factor-like 1; leucine zipper transcription factor-like protein 1; LZTFL1; wu:fb53f12; zgc:56268 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54585 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction