Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZTFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543167 Supplier Invitrogen™ Supplier No. PA543167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VQEQLHMAEK ELEKKFQQTA AYRNMKEILT KKNDQIKDLR KRLAQYEPED Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.

LZTFL1, also known as Leucine Zipper Transcription Factor-Like 1, is situated on the chromosomal region 3p21.3, which is frequently deleted in various cancers, including kidney cancer. It acts as a tumor suppressor by destabilizing AKT, a key protein in cell growth pathways, thus inhibiting tumor cell growth. Additionally, LZTFL1 has been implicated in the epithelial-mesenchymal transition in COVID-19, suggesting a role in viral response pathways. It is associated with Bardet-Biedl syndrome (BBS) and interacts with IFT27, a regulator of sperm flagellum formation, affecting spermatogenesis and male fertility. Knockout models show a phenotype of obesity, retinal degeneration, and abnormal cilia development, highlighting its role in normal ciliary function. The gene has two transcript isoforms and is part of a transcriptional map covering 250 kb, making it a subject of extensive genetic research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTFL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Lztfl1
Gene Accession No. Q9NQ48
Gene Alias 5530402H04Rik; 6130400H19Rik; AI414725; AW048545; BBS17; fb53f12; leucine zipper transcription factor like 1; leucine zipper transcription factor-like 1; leucine zipper transcription factor-like protein 1; LZTFL1; wu:fb53f12; zgc:56268
Gene Symbols Lztfl1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LZTFL1 (aa 250-299).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54585
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.