Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564381 Supplier Invitrogen™ Supplier No. PA564381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATVYSDKLWI FAGYDGNARL NDMWTIGLQD RELTCWEEVA QSGEIPPSCC NFPVAVCRDK MFVFSGQSGA KITNNLFQFE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

LZTR1, a member of the BTB-kelch superfamily, was initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Golgi complex. Deletion of this gene may be associated with DiGeorge syndrome, a developmental field defect involving the third and fourth pharyngeal pouches, causing the absence of thymus and parathyroid glands, congenital cardiac abnormalities and facial dysmorphism. LZTR1 is tyrosine phosphorylated and subsequently degraded upon induction of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LZTR1
Gene Accession No. Q8N653
Gene Alias 1200003E21Rik; AI591627; AW550890; BTBD29; fj87c05; leucine zipper like transcription regulator 1; leucine-zipper-like transcription regulator 1; leucine-zipper-like transcriptional regulator 1; leucine-zipper-like transcriptional regulator, 1; Lztr1; LZTR-1; MGC21205; NS10; si:dkey-98f17.2; SWNTS2; Tcfl2; wu:fj87c05; zgc:158250
Gene Symbols LZTR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LZTR1. Recombinant protein control fragment (Product #RP-104039).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8216
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.