Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZTR2 Polyclonal Antibody
LZTR2 Polyclonal Antibody
Supplier: Thermo Scientific PA589978
Description
LZTR2 Polyclonal Antibody for Western Blot

Specifications
LZTR2 | |
Polyclonal | |
Unconjugated | |
SEC16B | |
KIAA1928; Leucine zipper transcription regulator 2; LZTR2; PGPR-p117; protein SEC16 homolog B; protein transport protein Sec16B; Regucalcin gene promoter region-related protein p117; regucalcin gene promotor region related protein; RGPR; Rgpr-p117; SEC16 homolog B; SEC16 homolog B (S. cerevisiae); SEC16 homolog B, endoplasmic reticulum export factor; Sec16b; SEC16S | |
Rabbit | |
Affinity Chromatography | |
RUO | |
89866, 89867 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.75 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q91XT4, Q96JE7 | |
SEC16B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 250-550 of human SEC16B (NP_1491182). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction