Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZTR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520903
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520903 Supplier Invitrogen™ Supplier No. PA520903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse kidney tissue lysate. PA5-20903 can be used with blocking peptide PEP-1017.

LZTR2, also known as RGPR-p117, is a member of the BTB-kelch superfamily and was initially described as a nuclear factor I (NFI) binding protein and transcriptional regulator of the regucalcin gene. LZTR2 is cytoplasmically localized but is thought to translocate to the nucleus, a process mediated by protein kinase C signaling following hormonal stimulation. Recent evidence has suggested that there is a strong correlation of single nucleotide polymorphisms of LZTR2 with obesity in the Japanese population similar to that seen with the TMEM18 gene and the GNPDA2, BDNF, FAIM2, and MC4R genes with obesity in Caucasian populations, suggesting LZTR2 may play a role in metabolism and obesity risk.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTR2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SEC16B
Gene Accession No. Q75N33, Q91XT4, Q96JE7
Gene Alias KIAA1928; Leucine zipper transcription regulator 2; LZTR2; PGPR-p117; protein SEC16 homolog B; protein transport protein Sec16B; Regucalcin gene promoter region-related protein p117; regucalcin gene promotor region related protein; RGPR; Rgpr-p117; SEC16 homolog B; SEC16 homolog B (S. cerevisiae); SEC16 homolog B, endoplasmic reticulum export factor; Sec16b; SEC16S
Gene Symbols SEC16B
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human LZTR2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89866, 89867, 89868
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.