Learn More
Invitrogen™ LZTS1 Monoclonal Antibody (OTI3D12)

Description
LZTS1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LZTS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3D12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Lzts1 |
| Gene Accession No. | Q9Y250 |
| Gene Alias | F37; F37 esophageal cancer-related leucine zipper motif 1; F37/Esophageal cancer-related gene-coding leucine-zipper motif; FEZ1; leucine zipper putative tumor suppressor 1; leucine zipper tumor suppressor 1; leucine zipper, putative tumor suppressor 1; leucine-zipper protein FEZ1; LOW QUALITY PROTEIN: leucine zipper putative tumor suppressor 1; LZTS1; Psdzip70; PSD-Zip70 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.