Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZTS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541614 Supplier Invitrogen™ Supplier No. PA541614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QQSYVAMYQR NQRLEKALQQ LARGDSAGEP LEVDLEGADI PYEDIIATEI Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

This gene encodes a tumor suppressor protein that is ubiquitously expressed in normal tissues. In uveal melanomas, expression of this protein is silenced in rapidly metastasizing and metastatic tumor cells but has normal expression in slowly metastasizing or nonmetastasizing tumor cells. This protein may have a role in cell-cycle control by interacting with the Cdk1/cyclinB1 complex. This gene is located on chromosomal region 8p22. Loss of heterozygosity (LOH) in the 8p arm is a common characteristic of many types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTS1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Lzts1
Gene Accession No. Q9Y250
Gene Alias F37; F37 esophageal cancer-related leucine zipper motif 1; F37/Esophageal cancer-related gene-coding leucine-zipper motif; FEZ1; leucine zipper putative tumor suppressor 1; leucine zipper tumor suppressor 1; leucine zipper, putative tumor suppressor 1; leucine-zipper protein FEZ1; LOW QUALITY PROTEIN: leucine zipper putative tumor suppressor 1; LZTS1; Psdzip70; PSD-Zip70
Gene Symbols Lzts1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LZTS1 (aa 547-596).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11178
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.