Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZTS2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2410990.025MG
Description
LZTS2 Polyclonal specifically detects LZTS2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LZTS2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
LZTS2 | |
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human LZTS2. The immunogen is located within amino acids 570 - 620 of LZTS2. Amino Acid Squence: LQRERRRGEEQRDSFEG | |
0.025 mg | |
Cell Cycle and Replication | |
84445 | |
Human | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
hLZTS2, KIAA1813leucine zipper putative tumor suppressor 2, LAPSER1Protein LAPSER1, leucine zipper, putative tumor suppressor 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
LZTS2 antibody is predicted to not cross-react with other LZTS family members. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction