Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZTS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560871 Supplier Invitrogen™ Supplier No. PA560871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EARATLRVSE GRARGLQEAA RARELELEAC SQELQRHRQE AEQLREKAGQ LDAEAAGLRE PPVPPATADP FLLAESDEAK VQRAA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.

LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LZTS2
Gene Accession No. Q9BRK4
Gene Alias BC014695; HAMAP-Rule:MF_03026}; hLZTS2; KIAA1813; LAPSER1; Leucine zipper putative tumor suppressor 2; leucine zipper putative tumor suppressor 2 {ECO:0000255; leucine zipper tumor suppressor 2; leucine zipper, putative tumor suppressor 2; LOW QUALITY PROTEIN: leucine zipper putative tumor suppressor 2; Lzts2; mKIAA1813; Protein LAPSER1; putative tumor suppressor LAPSER1; RP11-108L7.8
Gene Symbols LZTS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LZTS2. Recombinant protein control fragment (Product #RP-95504).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84445
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.