Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ M 084 hydrochloride
GREENER_CHOICESDP

Catalog No. 580710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-071-0 10 mg
58-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-071-0 Supplier Tocris Bioscience™ Supplier No. 5807/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

TRPC4/5 channel blocker; antidepressant and anxiolytic

M 084 hydrochloride is a TRPC4/5 channel blocker (IC50 values are 3.7-10.3 and 8.2 μM, respectively). Also weakly blocks TRPC3 channels. Exhibits rapid antidepressant and anxiolytic effects in vivo.

Specifications

Chemical Name or Material N-Butyl-1H-benzimidazol-2-amine hydrochloride
CAS 1992047-63-8
Quantity 10 mg
Target TRPC Channel Blockers
Molecular Formula C11H15N3.HCl
Formula Weight 225.72
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.