Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Anisaldehyde, 97%

Catalog No. AC156761000
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 591-31-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-31-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key WMPDAIZRQDCGFH-UHFFFAOYSA-N
Synonym m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
PubChem CID 11569
IUPAC Name 3-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Density 1.1190g/mL
Boiling Point 143.0°C (50.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5540
Linear Formula CH3OC6H4CHO
Quantity 100 mL
Beilstein 08,59
Specific Gravity 1.119
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 136.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material m-Anisaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 209-712-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.