Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ M-cadherin Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50 - $596.50
Specifications
Antigen | M-cadherin |
---|---|
Concentration | 0.10 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110758
|
Invitrogen™
PA5110758 |
100 μL |
Each for $596.50
|
|
|||||
PIPA5111666
|
Invitrogen™
PA5111666 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: TFSARDPDTE QLQRLSYSKD YDPEDWLQVD AATGRIQTQH VLSPASPFLK GGWYRAIVLA QDDASQPRTA TGTLSIEILE VNDHAPVLAP PPPGSLCSEP HQGPGLLLGA TDEDLPPHGA PFHFQLSPRL PELGRNWSLS QVNVSH.
Cadherins are a multigene family of Ca++-dependent cell adhesion molecules. They are transmembrane glycoproteins consisting of an extracellular domain, which mediates Ca++-dependent intercellular adhesion by homophilic interactions, a transmembrane region and a cytoplasmic domain. The extracellular domain is divided into a series of subdomains designated EC1-EC5. Homolgies between different members of the cadherin family are most prominent in the cytoplasmic domain and in EC1 and EC2 and much less so in EC5 of the extracellular domain and in the transmembrane region. The binding properties and specificities of the adhesive function are located in the N-terminal part of the molecules. Four members of the cadherin family have been identified and molecularly cloned from mammalian cells. These include the neuronal (N), epithelial (E), placental (P) and muscle (M) cadherins. M-cadherin is not found in fibroblasts but is expressed at low level in myoblasts and is upregulated following induction of myotube formation, suggesting a specific function in skeletal muscle cell differentiation.Specifications
M-cadherin | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
CDH15 | |
AI323380; cadherin 15; cadherin 15, type 1, M-cadherin (myotubule); cadherin-14; Cadherin-15; cadherin-3; Cdh14; Cdh15; CDH3; CDHM; CRSBP-1; HAR; LYVE-1; M cadherin; Mcad; M-cadherin; MRD3; Muscle cadherin; muscle-cadherin; XLKD1 | |
CDH15 | |
Primary | |
Antigen Affinity Chromatography |
0.10 mg/mL | |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P55291 | |
1013 | |
Recombinant Protein corresonding to Human M-cadherin. Recombinant protein control fragment (Product #RP-90195). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title