Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol, 99%

Catalog No. AAA107330E
Click to view available options
Quantity:
250 g
1000 g
2500 g

CAS: 108-39-4 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.

Solubility
Miscible with water, diethylether, chloroform and ethanol.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym 3-Methylphenol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol

Specifications

Melting Point 11°C to 12°C
Boiling Point 202°C to 204°C
UN Number UN2076
Beilstein 506719
Merck Index 14,2579
Solubility Information Miscible with water,diethylether,chloroform and ethanol.
SMILES CC1=CC(=CC=C1)O
Formula Weight 108.14
Percent Purity 99%
Odor Phenol-like
Sensitivity Light sensitive
Density 1.034
pH 5
Quantity 2500 g
Flash Point 86°C (186°F)
Chemical Name or Material m-Cresol
Show More Show Less
Hazard Category H227-H301+H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, LIQUID
EINECSNumber 203-577-9
RTECSNumber GO6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.