Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
m-Cresol Purple TS, 0.1% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
CAS Min % | 98.0, 0.1 |
CAS Max % | 100.0, 0.1 |
Concentration | 0.1% (w/v) |
Percent Purity | 0.1% W/V |
Grade | Indicator |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Color | Red/Orange |
pH | 6.85 to 7.05 |
Quantity | 120 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction