Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSF Antibody (007), Alexa Fluor™ 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290794AF405
Description
M-CSF Monoclonal specifically detects M-CSF in Mouse samples. It is validated for ELISA.Specifications
| M-CSF | |
| Monoclonal | |
| Alexa Fluor 405 | |
| colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse M-CSF (P07141-1; Met1-Glu262). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers | |
| 1435 | |
| Mouse | |
| Purified |
| ELISA | |
| 7 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CSF1 Mouse Flt3-ligand | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction