Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSF Antibody (101), Alexa Fluor™ 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289985AF405
Description
M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Sandwich ELISA Detection.Specifications
M-CSF | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
101 | |
Sandwich ELISA Detection | |
colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human M-CSF (NP_757349.1; Met1-Asn190). | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers | |
1435 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction