Learn More
M-CSF Antibody (133), DyLight 755
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP290795IR
Catalog No. NB078060
Only null left
Add to Cart
Description
Description
M-CSF Monoclonal specifically detects M-CSF in Mouse samples. It is validated for ELISA.
Specifications
Specifications
M-CSF
Monoclonal
DyLight 755
colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse M-CSF (P07141-1; Met1-Glu262).
0.1 mL
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers
1435
Store at 4°C in the dark.
IgG
ELISA
133
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.