Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSF Antibody (OTI8E1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272601V
Description
M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
M-CSF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CSF1 | |
Recombinant protein fragment corresponding to amino acids 223-496 of human M-CSF (NP_000748) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI8E1 | |
Western Blot, Immunohistochemistry | |
colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers | |
1435.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction