Learn More
                                Invitrogen™ M-CSF Monoclonal Antibody (21113)
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$572.50 - $907.00
Specifications
| Antigen | M-CSF | 
|---|---|
| Clone | 21113 | 
| Concentration | 0.5 mg/mL | 
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles | 
| Applications | ELISA, Western Blot | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| PIMA523708 | Invitrogen™ MA523708 | 500 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $907.00
                                                 |  | |||||
| PIMA523813  | Invitrogen™ MA523813 | 100 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $572.50
                                                 |  | |||||
Description
This antibody detects recombinant human (rh)M-CSF in Western blots. In sandwich ELISAs, less than 7% cross-reactivity was observed with rmM-CSF. No significant cross-reactivity or interference was observed with over 75 other cytokines tested. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.Specifications
| M-CSF | |
| 0.5 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 5% trehalose and No Preservative | |
| P09603 | |
| 1435 | |
| E. coli-derived recombinant human M-CSF. | |
| Antibody | 
| 21113 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Lyophilized | |
| IgG2a | |
| Human | |
| Csf1 | |
| C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
| Csf1 | |
| Primary | |
| Protein A/G | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
