Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse M-CSF R/CD115 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB3818SP
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
M-CSFR/CD115 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P09581 | |
CSF1R | |
Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R/CD115 Ala20-Ser511 Accession # P09581 | |
25 μg | |
Primary | |
Detects mouse M-CSF R/CD115 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human M-CSF R/CD115, recombinant mouse (rm) PDGF R beta , or rmFlt-3 Ligand is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
460615 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1436 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction