Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSFR/CD115 Antibody (12-3A3-1B10), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143362AF647
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
M-CSFR/CD115 | |
Monoclonal | |
Alexa Fluor 647 | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
12-3A3-1B10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CSF1R | |
The immunogen for this antibody was CD115. | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
1436 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction