Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSFR/CD115 Antibody (61708), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB329C
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
M-CSFR/CD115 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
61708 | |
Flow Cytometry, CyTOF-ready | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115, Ile20-Glu512 (Pro54Ala), Accession # P07333.2 | |
0.5 mL | |
Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase | |
1436 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction