Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSFR/CD115 Antibody (AFS98), Biotin, Novus Biologicals™
SDP

Catalog No. p-5324330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14359205 0.05 mg
NBP14359201 0.1 mg
NBP1435925 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14359205 Supplier Novus Biologicals Supplier No. NBP1435920.05MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen M-CSF R/CD115
Applications Flow Cytometry
Classification Monoclonal
Clone AFS98
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Accession No. P07333
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Gene Symbols CSF1R
Host Species Rat
Immunogen The immunogen for this antibody was CD115.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1436
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.