Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCN1 Monoclonal Antibody (1)
GREENER_CHOICE

Catalog No. ABS0360102
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0360102 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0360102 Supplier Invitrogen™ Supplier No. ABS0360102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 036-01-02 detects M-Ficolin from human samples. ABS 036-01-02 has been successfully used in ELISA and IHC procedures. The ABS 036-01-02 immunogen is purified recombinant protein corresponding to residues 95-314 of human M-Ficolin, expressed in prokaryote. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

M-Ficolin (Ficolin-1) is a non-serum protein that is expressed in leukocytes and lung. M-Ficolin precipitates with mannose-binding lectin (MBL)-associated serine proteases (MASP)-1 and MASP-2, indicating that M-Ficolin forms complexes with MASP-1 and MASP-2, recognizes certain types of bacteria and subsequently activates the lectin pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCN1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene FCN1
Gene Accession No. O00602
Gene Alias collagen/fibrinogen domain-containing protein 1; FCN1; Fcna; FCNM; ficolin (collagen/fibrinogen domain containing) 1; ficolin 1; ficolin A; ficolin-1; Ficolin-A; ficolin-alpha; M-ficolin
Gene Symbols FCN1
Host Species Mouse
Immunogen Purified recombinant protein corresponding to residues 95-314 of human M-Ficolin, expressed in prokaryote.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2219
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.