Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCN1 Monoclonal Antibody (1E2B1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530594
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FCN1.
M-Ficolin (Ficolin-1) is a non-serum protein that is expressed in leukocytes and lung. M-Ficolin precipitates with mannose-binding lectin (MBL)-associated serine proteases (MASP)-1 and MASP-2, indicating that M-Ficolin forms complexes with MASP-1 and MASP-2, recognizes certain types of bacteria and subsequently activates the lectin pathway.
Specifications
FCN1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O00602 | |
FCN1 | |
Recombinant Human FCN1 protein (Met1-Ala326). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
1E2B1 | |
Unconjugated | |
FCN1 | |
collagen/fibrinogen domain-containing protein 1; FCN1; Fcna; FCNM; ficolin (collagen/fibrinogen domain containing) 1; ficolin 1; ficolin A; ficolin-1; Ficolin-A; ficolin-alpha; M-ficolin | |
Mouse | |
Protein A | |
RUO | |
2219 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction