Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551552 Supplier Invitrogen™ Supplier No. PA551552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGSSELRVDL VDFEGNHQFA KYKSFKVADE AEKYKLVLGA FVGGSAGNSL TGHNNNFFST KDQDNDVSSS NCAEKFQGAW WYADCHASNL NGLYLMGPHE SYANGINWS Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

M-Ficolin (Ficolin-1) is a non-serum protein that is expressed in leukocytes and lung. M-Ficolin precipitates with mannose-binding lectin (MBL)-associated serine proteases (MASP)-1 and MASP-2, indicating that M-Ficolin forms complexes with MASP-1 and MASP-2, recognizes certain types of bacteria and subsequently activates the lectin pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FCN1
Gene Accession No. O00602
Gene Alias collagen/fibrinogen domain-containing protein 1; FCN1; Fcna; FCNM; ficolin (collagen/fibrinogen domain containing) 1; ficolin 1; ficolin A; ficolin-1; Ficolin-A; ficolin-alpha; M-ficolin
Gene Symbols FCN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human M-Ficolin. Recombinant protein control fragment (Product #RP-93673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2219
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.