Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCN1 Recombinant Rabbit Monoclonal Antibody (017)

FCN1 Recombinant Rabbit Monoclonal Antibody (17)
Supplier: Thermo Scientific MA529270
Description
FCN1 Recombinant Monoclonal Antibody for IHC (P)

Specifications
FCN1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O00602 | |
FCN1 | |
Recombinant Human FCN1 protein (Met1-Ala326). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
017 | |
Unconjugated | |
FCN1 | |
collagen/fibrinogen domain-containing protein 1; FCN1; Fcna; FCNM; ficolin (collagen/fibrinogen domain containing) 1; ficolin 1; ficolin A; ficolin-1; Ficolin-A; ficolin-alpha; M-ficolin | |
Rabbit | |
Protein A | |
RUO | |
2219 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction