Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Nitrophenol, 99.9%, For GC analysis, MP Biomedicals™

Catalog No. ICN15589325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15589325 25 g
ICN15589305 5 g
ICN15589380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15589325 Supplier MP Biomedicals, Inc Supplier No. 0215589325
Add to Cart
Add to Cart

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Color Red-Yellow
Quantity 25 g
Formula Weight 139.11
Percent Purity 0.999
Physical Form Crystalline Powder
Chemical Name or Material m-Nitrophenol, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.