Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Phenylene Dibenzoate 99%, Thermo Scientific™

Catalog No. AC417491000
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-01-9
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00016576
InChI Key SUQGLJRNDJRARS-UHFFFAOYSA-N
Synonym 1,3-dibenzoyloxybenzene, resorcinol dibenzoate, 1,3-benzenediol, dibenzoate, 1,3-phenylene dibenzoate, m-phenylene dibenzoate, resorcinol, dibenzoate, 1,3-bis benzoyloxy benzene, 3-benzoyloxy phenyl benzoate, 1,3-benzenediol, 1,3-dibenzoate, 1,3-phenylenedibenzoate
PubChem CID 66742
IUPAC Name 3-(benzoyloxy)phenyl benzoate
SMILES O=C(OC1=CC(OC(=O)C2=CC=CC=C2)=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 117°C
Color Beige to White or Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 100 g
Beilstein 09, IV, 372
Formula Weight 318.32
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material m-Phenylene dibenzoate, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.