Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Phenylenediamine, +99%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
500g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
108-45-2
C6H8N2
108.14
MFCD00007799
WZCQRUWWHSTZEM-UHFFFAOYSA-N
m-phenylenediamine,1,3-phenylenediamine,1,3-diaminobenzene,1,3-benzenediamine,m-diaminobenzene,3-aminoaniline,m-aminoaniline,meta-phenylenediamine,developer c,3-phenylenediamine
7935
CHEBI:8092
benzene-1,3-diamine
C1=CC(=CC(=C1)N)N
108-45-2
C6H8N2
108.14
MFCD00007799
WZCQRUWWHSTZEM-UHFFFAOYSA-N
m-phenylenediamine,1,3-phenylenediamine,1,3-diaminobenzene,1,3-benzenediamine,m-diaminobenzene,3-aminoaniline,m-aminoaniline,meta-phenylenediamine,developer c,3-phenylenediamine
7935
CHEBI:8092
benzene-1,3-diamine
C1=CC(=CC(=C1)N)N
Packaging Glass bottle
Quantity 25g
Flash Point 175°C
Color Brown-Gray or Tan to Yellow
Infrared Spectrum Authentic
Boiling Point 282.0°C to 284.0°C
Melting Point 63.0°C to 65.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H4(NH2)2
Beilstein 03, IV, 79
Merck Index 15, 7395
Solubility Information Solubility in water: 350g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 108.14
Physical Form Pellets
Percent Purity ≥99%
Chemical Name or Material m-Phenylenediamine
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : this product tends to darken during storage