Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-Ras/R-Ras3 Antibody (26E12), Novus Biologicals™

Mouse Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | M-Ras/R-Ras3 |
---|---|
Clone | 26E12 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
M-Ras/R-Ras3 | |
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
Recombinant Human M-Ras/R-Ras3 protein [UniProt O14807] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
26E12 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Signal Transduction | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
22808 | |
IgG2a | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title