Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-Ras/R-Ras3 Antibody (OTI3C4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272776IR
Description
M-Ras/R-Ras3 Monoclonal specifically detects M-Ras/R-Ras3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
M-Ras/R-Ras3 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MRAS | |
Full length recombinant protein of human M-Ras/R-Ras3 (NP_036351) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3C4 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
22808.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction