Learn More
m-Toluenesulfonyl chloride, 98%
Description
Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
Specifications
Specifications
| Density | 1.314 |
| Boiling Point | 252°C to 253°C |
| Flash Point | 107°C (224°F) |
| Odor | Acrid |
| Refractive Index | 1.548 |
| Quantity | 100 g |
| UN Number | UN3265 |
| Beilstein | 2087865 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.