Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Toluidine, 99%

Catalog No. p-7021646
Click to view available options
Quantity:
250 g
1000 g

CAS: 108-44-1 | C7H9N | 107.16 g/mol

m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

Solubility
Soluble in water (2g/L), (10 g/L at 25°C ).

Notes
Keep container tightly sealed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-44-1
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00007808
InChI Key JJYPMNFTHPTTDI-UHFFFAOYSA-N
Synonym m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine
PubChem CID 7934
IUPAC Name 3-methylaniline
SMILES CC1=CC=CC(N)=C1

Specifications

Melting Point -30°C
Density 0.992
Boiling Point 203°C to 204°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.567
Quantity 250 g
UN Number UN1708
Beilstein 635944
Merck Index 14,9536
Solubility Information Soluble in water (2g/L),(10g/L at 25°C ).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material m-Toluidine
Show More Show Less
Hazard Category H227-H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P374-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID
EINECSNumber 203-583-1
RTECSNumber XU2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.