Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Toluidine, 99%

Catalog No. ac157870050
Click to view available options
Quantity:
5 mL
1 L
5 L
Packaging:
Glass Bottle
Plastic drum

CAS: 108-44-1 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-44-1
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00007808
InChI Key JJYPMNFTHPTTDI-UHFFFAOYSA-N
Synonym m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine
PubChem CID 7934
IUPAC Name 3-methylaniline
SMILES CC1=CC=CC(N)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color White
Density 0.9800g/mL
Boiling Point 203.0°C to 204.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5660 to 1.5680
Linear Formula CH3C6H4NH2
Quantity 5 mL
Beilstein 12,853
Merck Index 15,9696
Specific Gravity 0.98
Solubility Information Solubility in water: 0.2 g/100mL (20°C). Other solubilities: >50 g/100mL methanol, >50 g/100mL ethanol, soluble in ether, dmf and diluted acids
Formula Weight 107.15
Percent Purity 99%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material m-Toluidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 203-583-1
RTECSNumber XU2800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.