Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolylacetic acid, 99%

Catalog No. AAA1774318
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1774318 50 g
AAA1774309 10 g
AAA1774330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1774318 Supplier Thermo Scientific Chemicals Supplier No. A1774318
Add to Cart

CAS: 621-36-3 | C9H10O2 | 150.18 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-36-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00004340
InChI Key GJMPSRSMBJLKKB-UHFFFAOYSA-N
Synonym 3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate
PubChem CID 12121
IUPAC Name 2-(3-methylphenyl)acetic acid
SMILES CC1=CC=CC(CC(O)=O)=C1

Specifications

Melting Point 62°C to 66°C
Quantity 50 g
Beilstein 2043549
Solubility Information Soluble in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material m-Tolylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-683-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.