Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene (Synthesis Grade), Thermo Scientific™

Catalog No. ac18086n219 Shop All Fisherbrand Products
Click to view available options
Quantity:
19 L

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -48°C
Color Colorless
Density 0.86g/mL
Boiling Point 139°C
Quantity 19 L
Flash Point 25°C (77°F)
Physical Form Liquid
Linear Formula C6H4(CH3)2
Vapor Pressure 8mbar at 20°C
Viscosity 0.62mPa/s at 20°C
Formula Weight 106.16
Grade Synthesis
Chemical Name or Material m-Xylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.