Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene, EMD-Millipore

Catalog No. m1097970005 Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mL

Hydrocarbons C8

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym 1, 3-Dimethylbenzene
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Packaging Glass ampoule
Explosion Limit 1.1 to 7 %(V)
Ignition Point 525°C
Vapor Pressure 8 hPa (20°C)
Density Density: 0.86g/cm3 (20°)
Assay Percent Range ≥99.3%
Linear Formula C6H4(CH3)2
Quantity 5 mL
Formula Weight 106.16
Chemical Name or Material m-Xylene
Recommended Storage Store without restrictions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.