Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylene dibromide, 97%

Catalog No. p-7027898
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 626-15-3 | C8H8Br2 | 263.96 g/mol

It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, active metals, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-15-3
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000178
InChI Key OXHOPZLBSSTTBU-UHFFFAOYSA-N
Synonym 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene
PubChem CID 69373
IUPAC Name 1,3-bis(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)CBr)CBr

Specifications

Melting Point 74°C to 76°C
Quantity 10 g
UN Number UN3261
Beilstein 971085
Solubility Information Insoluble in water.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material m-Xylylene dibromide
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-931-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.