Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylene dichloride, 97%

Catalog No. p-7081534
Click to view available options
Quantity:
25 g
100 g

CAS: 626-16-4 | C8H8Cl2 | 175.052 g/mol

m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-16-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000912
InChI Key GRJWOKACBGZOKT-UHFFFAOYSA-N
Synonym 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro
PubChem CID 12275
IUPAC Name 1,3-bis(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)CCl)CCl

Specifications

Melting Point 33°C to 37°C
Density 1.302
Boiling Point 250°C to 255°C
Flash Point 144°C (291°F)
Quantity 25 g
UN Number UN2928
Beilstein 2079898
Solubility Information Insoluble in water.
Formula Weight 175.06
Percent Purity 97%
Chemical Name or Material m-Xylylene dichloride
Show More Show Less
Hazard Category H302-H314-H330
Hazard Statement GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 210-932-1
RTECSNumber ZE4070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.