Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13 bacteriophage (g3p) Mouse anti-Bacteria, Biotin, Clone: E1, Exalpha
SDP

Catalog No. 502102310
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-2310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-2310 Supplier Vector Laboratories Supplier No. Z110M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

M13 bacteriophage (g3p) Monoclonal antibody specifically detects M13 bacteriophage (g3p) in Bacteria samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Specifications

Antigen M13 bacteriophage (g3p)
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone E1
Concentration 0.5-2 mg/mL
Conjugate Biotin
Formulation PBS with 0.08% sodium azide; pH
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with isolated M13 phage coat proteins and mouse myeloma cells.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.