Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M6PR Monoclonal Antibody (1B11C1)
GREENER_CHOICE

Catalog No. FERMA548539
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548539 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548539 Supplier Invitrogen™ Supplier No. MA548539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

M6PR Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

M6PR is a receptor for mannose-6-phosphate groups on lysosomal enzymes. The receptor forms a homodimer or homotetramer for intracellular targeting of lysosomal enzymes and export of newly synthesized lysosomal enzymes into the cell secretions. The receptor is an integral membrane protein which localizes to the trans-Golgi reticulum, endosomes, and the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M6PR
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 1B11C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene M6PR
Gene Accession No. P20645, P24668
Gene Alias 46 kDa mannose 6-phosphate receptor; 46-kDa mannose 6-phosphate receptor; 46mpr; Cation-dependent mannose-6-phosphate receptor; CD Man-6-P receptor; CD-M6PR; CD-MPR; FLJ32994; M6pr; mannose-6-phosphate receptor (cation dependent); mannose-6-phosphate receptor, cation dependent; MPR 46; MPR46; MPR-46; MPRD; Mr 46,000 Man6PR; small mannose 6-phosphate receptor; SMPR
Gene Symbols M6PR
Host Species Mouse
Immunogen Purified recombinant fragment of human M6PR (AA: 124-277) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17113, 4074
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.