Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ M6PR Monoclonal Antibody (3A10B2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548540
Description
M6PR Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
M6PR is a receptor for mannose-6-phosphate groups on lysosomal enzymes. The receptor forms a homodimer or homotetramer for intracellular targeting of lysosomal enzymes and export of newly synthesized lysosomal enzymes into the cell secretions. The receptor is an integral membrane protein which localizes to the trans-Golgi reticulum, endosomes, and the plasma membrane.
Specifications
M6PR | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P20645, P24668 | |
M6PR | |
Purified recombinant fragment of human M6PR (AA: 124-277) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
3A10B2 | |
Unconjugated | |
M6PR | |
46 kDa mannose 6-phosphate receptor; 46-kDa mannose 6-phosphate receptor; 46mpr; Cation-dependent mannose-6-phosphate receptor; CD Man-6-P receptor; CD-M6PR; CD-MPR; FLJ32994; M6pr; mannose-6-phosphate receptor (cation dependent); mannose-6-phosphate receptor, cation dependent; MPR 46; MPR46; MPR-46; MPRD; Mr 46,000 Man6PR; small mannose 6-phosphate receptor; SMPR | |
Mouse | |
Protein G | |
RUO | |
17113, 4074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction