Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M6PR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592876 Supplier Invitrogen™ Supplier No. PA592876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEEKTCDLVG EKGKESEKEL ALVKRLKPLF NKSFESTVGQ GSDTYIYIFR VCREAGNHTS GAGLVQINKS NGKETVVGRL NETHIFNGSN WIMLIYKGGD EYDNHCGKEQ RRAVVMISCN RHTLADNFNP VSEERGKVQD CFYLFEMDSS LACSPEISH; Positive Samples: Mouse kidney; Cellular Location: Lysosome membrane, Single-pass type I membrane protein.

M6PR is a receptor for mannose-6-phosphate groups on lysosomal enzymes. The receptor forms a homodimer or homotetramer for intracellular targeting of lysosomal enzymes and export of newly synthesized lysosomal enzymes into the cell secretions. The receptor is an integral membrane protein which localizes to the trans-Golgi reticulum, endosomes, and the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M6PR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene M6PR
Gene Accession No. P20645, P24668, Q6AY20
Gene Alias 46 kDa mannose 6-phosphate receptor; 46-kDa mannose 6-phosphate receptor; 46mpr; Cation-dependent mannose-6-phosphate receptor; CD Man-6-P receptor; CD-M6PR; CD-MPR; FLJ32994; M6pr; mannose-6-phosphate receptor (cation dependent); mannose-6-phosphate receptor, cation dependent; MPR 46; MPR46; MPR-46; MPRD; Mr 46,000 Man6PR; small mannose 6-phosphate receptor; SMPR
Gene Symbols M6PR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-185 of human M6PR (NP_0023461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17113, 312689, 4074
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.