Learn More
Invitrogen™ M6PR Polyclonal Antibody

Description
Antibody detects endogenous levels of total M6PR.
Specifications
Specifications
| Antigen | M6PR |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | M6PR |
| Gene Accession No. | P20645, P24668, Q6AY20 |
| Gene Alias | 46 kDa mannose 6-phosphate receptor; 46-kDa mannose 6-phosphate receptor; 46mpr; Cation-dependent mannose-6-phosphate receptor; CD Man-6-P receptor; CD-M6PR; CD-MPR; FLJ32994; M6pr; mannose-6-phosphate receptor (cation dependent); mannose-6-phosphate receptor, cation dependent; MPR 46; MPR46; MPR-46; MPRD; Mr 46,000 Man6PR; small mannose 6-phosphate receptor; SMPR |
| Gene Symbols | M6PR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.