Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M6PR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111123 Supplier Invitrogen™ Supplier No. PA5111123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKRLKPLFNK SFESTVGQGS DTYIYIFRVC REAGNHTSGA GLVQINKSNG KETVVGRLNE THIFNGSNWI MLIYKGGDEY DNHCGKEQRR AVVMISCNRH TLADNFNPVS EERGKVQDCF YLFE Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.

M6PR is a receptor for mannose-6-phosphate groups on lysosomal enzymes. The receptor forms a homodimer or homotetramer for intracellular targeting of lysosomal enzymes and export of newly synthesized lysosomal enzymes into the cell secretions. The receptor is an integral membrane protein which localizes to the trans-Golgi reticulum, endosomes, and the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M6PR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene M6PR
Gene Accession No. P20645
Gene Alias 46 kDa mannose 6-phosphate receptor; 46-kDa mannose 6-phosphate receptor; 46mpr; Cation-dependent mannose-6-phosphate receptor; CD Man-6-P receptor; CD-M6PR; CD-MPR; FLJ32994; M6pr; mannose-6-phosphate receptor (cation dependent); mannose-6-phosphate receptor, cation dependent; MPR 46; MPR46; MPR-46; MPRD; Mr 46,000 Man6PR; small mannose 6-phosphate receptor; SMPR
Gene Symbols M6PR
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human M6PR. Recombinant protein control fragment (Product #RP-102892).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.