Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ M8 B hydrochloride
GREENER_CHOICE

Catalog No. 532450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-245-0 50 mg
53-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-245-0 Supplier Tocris Bioscience™ Supplier No. 5324/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective TRPM8 channel blocker

M8 B hydrochloride is a potent and selective TRPM8 channel blocker. Blocks cold- and icilin -induced TRPM8 activation in vitro (IC50 values are 7.8 and 26.9 nM respectively). Exhibits no effect at other TRP channels (IC50 > 20 μM). Reduces body temperature in vivo.

Specifications

Quantity 50 mg
Product Type M8 B hydrochloride
Purity >99%
Molecular Weight (g/mol) 432.96
Inhibitors M8 B hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.