Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAB21L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566289 Supplier Invitrogen™ Supplier No. PA566289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPTEFEVVLYL NQMGVFNFVD DGSLPGCAVL KLSDGRKRSM SLWVEFITAS GYLSARKIRS RFQTLVAQAV DKCSYRDVVK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene is similar to the MAB-21 cell fate-determining gene found in C. elegans. It may be involved in eye and cerebellum development, and it has been proposed that expansion of a trinucleotide repeat region in the 5' UTR may play a role in a variety of psychiatric disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAB21L1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAB21L1
Gene Accession No. Q13394
Gene Alias AW047968; CAGR1; Mab21; mab-21 like 1; MAB21L1; mab-21-like 1 (C. elegans); mab-21-like protein 1; mb211; Nbla00126; protein mab-21-like 1; Putative nucleotidyltransferase MAB21L1
Gene Symbols MAB21L1
Host Species Rabbit
Immunogen Recombinant Human MAB21L1. Recombinant protein control fragment (Product #RP-105636).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.